1,4-Dibromonaphthalene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18171-06
A18171-14
AAA18171-06EA
131
USD
AAA18171-06
AAA18171-14
1,4-Dibromonaphthalene 98%
1,4-Dibromonaphthalene
Purity: 98%
CAS No.: 83-53-4
Molecular Formula: C10H6Br2
Formula Weight: 285.97
Melting Point: 80-82°
MDL No.: MFCD00041823
Formula:
C₁₀H₆Br₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041823 CAS Number: 83-53-4 EINECS: 201-484-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...