D-Cycloserine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Inhibits bacterial cell wall biosynthesis. Prevents conversion of D-Alanine to L-Alanine
Purity: 98+%
CAS No.: 68-41-7
Molecular Formula: C3H6N2O2
Formula Weight: 102.09
Storage Temperature: 2°C to 8°C
Melting Point: 137° dec.
Beilstein Reference: 80798
Merck Reference: 14,2751
Optical Rotation: +115° (c=2 in water)
MDL No.: MFCD00005353
Formula:
C₃H₆N₂O₂ Melting Pt: 137 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005353 CAS Number: 68-41-7 EINECS: 200-688-4 Merck Index: 14,2751 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...