D-(-)-Ribose 98%
Supplier: Thermo Scientific Chemicals
A17894-09
A17894-30
A17894-18
AAA17894-09EA
29.89
USD
AAA17894-09
AAA17894-30
AAA17894-18
D-(-)-Ribose 98%
D(-)-Ribose
Purity: 98%
CAS No.: 50-69-1
Molecular Formula: C5H10O5
Formula Weight: 150.13
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 82-86°
Beilstein Reference: 1723081
Merck Reference: 14,8204
Optical Rotation: -20° (c=10 in water, 28h)
MDL No.: MFCD00135453
Formula:
C₅H₁₀O₅ Boiling Pt: 331 °C (1013 hPa) Melting Pt: 82…86 °C Density: 0.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00135453 CAS Number: 50-69-1 EINECS: 200-059-4 Merck Index: 14,8204 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...