5-Nitro-o-toluidine 98+%

Supplier: Thermo Scientific Chemicals
Danger

A17726-36 A17726-0E A17726-22
AAA17726-36EA 164 USD
AAA17726-36 AAA17726-0E AAA17726-22
5-Nitro-o-toluidine 98+%
5-Nitro-o-toluidine

Purity: 98+% CAS No.: 99-55-8 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Odor: Characteristic Melting Point: 105-108° Beilstein Reference: 879021 UN No.: 2660 MDL No.: MFCD00007741

Formula: C₇H₈N₂O₂
Melting Pt: 105…108 °C
Density: 1.365 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007741
CAS Number: 99-55-8
EINECS: 202-765-8
UN: 2660
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR