D-(-)-Fructose 99%
Supplier: Thermo Scientific Chemicals
A17718-0I
A17718-0B
A17718-30
AAA17718-0IEA
169.44
USD
AAA17718-0I
AAA17718-0B
AAA17718-30
D-(-)-Fructose 99%
D(-)-Fructose
Purity: 99%
CAS No.: 57-48-7
Molecular Formula: C6H12O6
Formula Weight: 180.16
Odor: Odorless
Melting Point: ca 105-110°
Beilstein Reference: 1239004
Merck Reference: 14,4273
Optical Rotation: -89° (c=2 in water)
MDL No.: MFCD00148910
Formula:
C₆H₁₂O₆ Boiling Pt: 552 °C (1013 hPa) Melting Pt: ∼ 105…110 °C Density: 1.59 Storage Temperature: Ambient |
MDL Number:
MFCD00148910 CAS Number: 57-48-7 EINECS: 200-333-3 Merck Index: 14,4273 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...