D-(-)-Fructose 99%

Supplier: Thermo Scientific Chemicals

A17718-0I A17718-0B A17718-30
AAA17718-0IEA 169.44 USD
AAA17718-0I AAA17718-0B AAA17718-30
D-(-)-Fructose 99%
D(-)-Fructose

Purity: 99% CAS No.: 57-48-7 Molecular Formula: C6H12O6 Formula Weight: 180.16 Odor: Odorless Melting Point: ca 105-110° Beilstein Reference: 1239004 Merck Reference: 14,4273 Optical Rotation: -89° (c=2 in water) MDL No.: MFCD00148910

Formula: C₆H₁₂O₆
Boiling Pt: 552 °C (1013 hPa)
Melting Pt: ∼ 105…110 °C
Density: 1.59
Storage Temperature: Ambient
MDL Number: MFCD00148910
CAS Number: 57-48-7
EINECS: 200-333-3
Merck Index: 14,4273
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c96c34b23753d10b06108938876644c747995c&vid=6d952afc3154d3befcf243a015f513a9130c0835&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR