2-Nitroethanol tech. 80%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

A17681-14 A17681-22 A17681-06
AAA17681-14EA 171 USD
AAA17681-14 AAA17681-22 AAA17681-06
2-Nitroethanol tech. 80%, Technical Grade
2-Nitroethanol

Purity: 80% CAS No.: 625-48-9 Molecular Formula: O2NCH2CH2OH Formula Weight: 91.07 Flash Point: 110°(230°F) Refractive Index: 1.4451 Melting Point: -80° Boiling Point: 194° Beilstein Reference: 1633753 MDL No.: MFCD00007405

Formula: C₂H₅NO₃
MW: 91.07 g/mol
Boiling Pt: 194
Melting Pt: -80
Density: 1.270
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007405
CAS Number: 625-48-9
EINECS: 210-895-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR