Dexamethasone 98%

Supplier: Thermo Scientific Chemicals
Warning

A17590-03 A17590-14 A17590-06
AAA17590-03EA 175.55 USD
AAA17590-03 AAA17590-14 AAA17590-06
Dexamethasone 98%
Dexamethasone

Anti-inflammatory glucocorticoid


Purity: 98% CAS No.: 50-02-2 Molecular Formula: C22H29FO5 Formula Weight: 392.47 Melting Point: ca 260° dec. Beilstein Reference: 2066651 Merck Reference: 14,2943 Optical Rotation: +75° (c=1 in doxane) MDL No.: MFCD00064136


Anti-inflammatory glucocorticoid

Formula: C₂₂H₂₉FO₅
Melting Pt: ca 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064136
CAS Number: 50-02-2
EINECS: 200-003-9
Merck Index: 14,2943

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR