n-Dotriacontane 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Dotriacontane

A17480-14 A17480-06
AAA17480-14EA 150 USD
AAA17480-14 AAA17480-06
n-Dotriacontane 98%
n-Dotriacontane

Purity: 98% CAS No.: 544-85-4 Molecular Formula: CH3(CH2)30CH3 Formula Weight: 450.88 Melting Point: 68-71° Boiling Point: 300-310° MDL No.: MFCD00009411

Formula: H₃C(CH₂)₃₀CH₃
Boiling Pt: 300…310 °C
Melting Pt: 68…71 °C
Density: 0.812
Storage Temperature: Ambient
MDL Number: MFCD00009411
CAS Number: 544-85-4
EINECS: 208-881-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR