Tetramethylthiuram monosulfide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(dimethylthiocarbamyl) sulphide, Bis(dimethylthiocarbamyl) sulfide

A17259-22
AAA17259-22EA 67.6 USD
AAA17259-22 AAA17259-36
Tetramethylthiuram monosulfide 97%
Tetramethylthiuram monosulphide

Purity: 97% CAS No.: 97-74-5 Molecular Formula: C6H12N2S3 Formula Weight: 208.37 Melting Point: 106-110° Beilstein Reference: 1775650

Formula: C₆H₁₂N₂S₃
Melting Pt: 106…110 °C
Density: 1.37
Storage Temperature: Ambient
MDL Number: MFCD00014870
CAS Number: 97-74-5
EINECS: 202-605-7
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR