N-(1-Naphthyl)ethylenediamine dihydrochloride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17164-18
A17164-09
AAA17164-18EA
238.42
USD
AAA17164-18
AAA17164-09
N-(1-Naphthyl)ethylenediamine dihydrochloride 96%
N-2-Aminoethyl-1-naphthylamine dihydrochloride
Purity: 96%
CAS No.: 1465-25-4
Molecular Formula: C12H14N2.2HCl
Formula Weight: 259.18
Melting Point: 191-199°
Beilstein Reference: 3707471
Merck Reference: 14,6409
MDL No.: MFCD00012556
Formula:
C₁₂H₁₆Cl₂N₂ Melting Pt: 191…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012556 CAS Number: 1465-25-4 EINECS: 215-981-2 Merck Index: 14,6409 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...