2-Chloronicotinic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid

A17082-09 A17082-18 A17082-30
AAA17082-09EA 28.9 USD
AAA17082-09 AAA17082-18 AAA17082-30
2-Chloronicotinic acid 99%
2-Chloronicotinic acid

Purity: 99% CAS No.: 2942-59-8 Molecular Formula: C6H4ClNO2 Formula Weight: 157.56 Melting Point: ca 180° dec. Beilstein Reference: 119023

Formula: C₆H₄ClNO₂
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: ∼ 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006236
CAS Number: 2942-59-8
EINECS: 220-937-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR