Xylitol 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
A16944-22
A16944-36
AAA16944-22EA
59.76
USD
AAA16944-22
AAA16944-36
Xylitol 99%
Xylitol
Purity: 99%
CAS No.: 87-99-0
Molecular Formula: C5H12O5
Formula Weight: 152.15
Odor: Odorless
Melting Point: 92-96°
Beilstein Reference: 1720523
Merck Reference: 14,10085
MDL No.: MFCD00064292
Formula:
C₅H₁₂O₅ Boiling Pt: 495 °C (1013 hPa) Melting Pt: 92…96 °C Density: 1.515 Storage Temperature: Ambient |
MDL Number:
MFCD00064292 CAS Number: 87-99-0 EINECS: 201-788-0 Merck Index: 14,10085 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...