(R)-(+)-1-(1-Naphthyl)ethylamine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine

A16918-14 A16918-06
AAA16918-14EA 477 USD
AAA16918-14 AAA16918-06
(R)-(+)-1-(1-Naphthyl)ethylamine 99%
(R)-(+)-1-(1-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
Boiling Pt: 152…153 °C/11 mm
Density: 1.060
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00064114
CAS Number: 3886-70-2
EINECS: 223-425-5
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR