Alizarin complexone, (max. 10% H₂O)

Supplier: Thermo Scientific Chemicals

A16699-03 A16699-06
AAA16699-03EA 97.02 USD
AAA16699-03 AAA16699-06
Alizarin complexone, (max. 10% H₂O)
Alizarin complexone

CAS No.: 3952-78-1 Molecular Formula: C19H15NO8 Formula Weight: 385.33 Melting Point: 180-182° Beilstein Reference: 2190028 MDL No.: MFCD00001202

Formula: C₁₉H₁₅NO₈
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00001202
CAS Number: 3952-78-1
EINECS: 223-544-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR