7-Hydroxy-4-(trifluoromethyl)coumarin 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Trifluoromethyl)umbelliferone, TFMU
A16537-03
A16537-06
AAA16537-03EA
70.5
USD
AAA16537-03
AAA16537-06
7-Hydroxy-4-(trifluoromethyl)coumarin 98%
7-Hydroxy-4-(trifluoromethyl)coumarin
Purity: 98%
CAS No.: 575-03-1
Molecular Formula: C10H5F3O3
Formula Weight: 230.14
Melting Point: 178-180°
MDL No.: MFCD00037578
Formula:
C₁₀H₅F₃O₃ Melting Pt: 178…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037578 CAS Number: 575-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...