Fast green FCF
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stain for showing collagen, muscle, cytoplasm and cells in mammalian tissues
CAS No.: 2353-45-9
Molecular Formula: C37H34N2Na2O10S3
Formula Weight: 808.86
Melting Point: ca 290° dec.
Beilstein Reference: 5718212
Merck Reference: 14,3941
MDL No.: MFCD00013053
Formula:
C₃₇H₃₄N₂Na₂O₁₀S₃ Melting Pt: ∼ 290 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013053 CAS Number: 2353-45-9 EINECS: 219-091-5 Merck Index: 14,3941 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...