Benzethonium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hyamine® 1622, N-Benzyl-N,N-dimethyl-2-(2-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride
A16142-22
AAA16142-22EA
84.01
USD
AAA16142-22
AAA16142-0E
Benzethonium chloride 97%
Benzethonium chloride
MDL: MFCD00011742
Beilstein: 3898548
Soluble in water
Purity: 97%
CAS No.: 121-54-0
Molecular Formula: C27H42ClNO2
Formula Weight: 448.10
Odor: Odorless
Melting Point: 157-165°
Beilstein Reference: 3898548
Merck Reference: 14,1074
UN No.: 1759
MDL No.: MFCD00011742
Formula:
C₂₇H₄₂ClNO₂ MW: 448.09 g/mol Melting Pt: 157…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011742 CAS Number: 121-54-0 EINECS: 204-479-9 UN: 2923 ADR: 8,III Merck Index: 14,1074 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...