Methylcyclohexane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99%
CAS No.: 108-87-2
Molecular Formula: C7H14
Formula Weight: 98.19
Flash Point: -4°(-25°F)
Refractive Index: 1.4230
Melting Point: -126°
Boiling Point: 101°
Beilstein Reference: 505972
Merck Reference: 14,6047
UN No.: 2296
MDL No.: MFCD00001497
Formula:
C₆H₁₁CH₃ Boiling Pt: 101 °C Melting Pt: –126 °C Density: 0.771 Flash Pt: –4 °C (–25 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001497 CAS Number: 108-87-2 EINECS: 203-624-3 UN: 2296 ADR: 3,II Merck Index: 14,6047 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...