Methylcyclohexane 99%
Supplier: Thermo Scientific Chemicals
Purity: 99%
CAS No.: 108-87-2
Molecular Formula: C7H14
Formula Weight: 98.19
Flash Point: -4°(-25°F)
Refractive Index: 1.4230
Melting Point: -126°
Boiling Point: 101°
Beilstein Reference: 505972
Merck Reference: 14,6047
UN No.: 2296
MDL No.: MFCD00001497
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...