1-Decanethiol 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: n-Decanethiol, 1-Mercaptodecane, n-Decyl mercaptan

A15913-AC A15913-AE A15913-AP
AAAA15913-ACEA 51.33 USD
AAAA15913-AC AAA15913-AE AAA15913-AP
1-Decanethiol 96%
1-Decanethiol

Purity: 96% CAS No.: 143-10-2 Molecular Formula: C10H22S Formula Weight: 174.35 Flash Point: 98°(208°F) Refractive Index: 1.4565 Odor: Unpleasant Melting Point: -26° Boiling Point: 237° Beilstein Reference: 1735223 MDL No.: MFCD00004884

Formula: C₁₀H₂₂S
Boiling Pt: 237 °C
Melting Pt: –26 °C
Density: 0.843
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004884
CAS Number: 143-10-2
EINECS: 205-584-2
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR