Sulfanilic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Reagent for the detection of histidine; intermediate for dyes and pharmaceuticals
Purity: 98+%
CAS No.: 121-57-3
Molecular Formula: C6H7NO3S
Formula Weight: 173.19
Physical Form: Cryst powder
Density: 1.485
Appearance: White to pale brown
Melting Point: ca 365°
Beilstein Reference: 908765
Merck Reference: 14,8926
MDL No.: MFCD00007886
Reagent for the detection of histidine; intermediate for dyes and pharmaceuticals
Formula:
4-(H₂N)C₆H₄SO₃H Boiling Pt: 363 °C (1013 hPa) Melting Pt: ∼ 365 °C Density: 1.485 Storage Temperature: Ambient |
MDL Number:
MFCD00007886 CAS Number: 121-57-3 EINECS: 204-482-5 Merck Index: 14,8926 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...