Quercetin dihydrate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15807-14
A15807-22
A15807-36
AAA15807-14EA
70.2
USD
AAA15807-14
AAA15807-22
AAA15807-36
Quercetin dihydrate 97%
Quercetin dihydrate
Inhibitor of protein tyrosine kinase
Purity: 97%
CAS No.: 6151-25-3
Molecular Formula: C15H10O7.2H2O
Formula Weight: 338.28 (302.25anhy)
Odor: Odorless
Melting Point: 310-315°
Beilstein Reference: 317313
UN No.: 2811
MDL No.: MFCD00149487
Applications: Inhibitor of protein tyrosine kinase
Formula:
C₁₅H₁₄O₉ Melting Pt: 310…315 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149487 CAS Number: 6151-25-3 EINECS: 204-187-1 UN: 2811 ADR: "6.1",III Merck Index: 14,8034 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...