Sodium chlorodifluoroacetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15756-22
A15756-36
A15756-14
AAA15756-22EA
236
USD
AAA15756-22
AAA15756-36
AAA15756-14
Sodium chlorodifluoroacetate 97%
Sodium chlorodifluoroacetate
Purity: 97%
CAS No.: 1895-39-2
Molecular Formula: ClCF2CO2Na
Formula Weight: 152.46
Melting Point: ca 195° dec.
Beilstein Reference: 4162363
Formula:
C₂ClF₂NaO₂ Melting Pt: ∼ 195 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064771 CAS Number: 1895-39-2 EINECS: 217-586-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...