2-Amino-5-chlorobenzophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15720-0E
A15720-36
A15720-22
AAA15720-0EEA
448
USD
AAA15720-0E
AAA15720-36
AAA15720-22
2-Amino-5-chlorobenzophenone 98+%
2-Amino-5-chlorobenzophenone
Purity: 98+%
CAS No.: 719-59-5
Molecular Formula: C13H10ClNO
Formula Weight: 231.68
Flash Point: 211°(412°F)
Melting Point: 96-99°
Beilstein Reference: 475640
MDL No.: MFCD00007839
Formula:
C₁₃H₁₀ClNO Boiling Pt: 424 °C (1013 hPa) Melting Pt: 96…99 °C Density: 1,33 Flash Pt: 211 °C(412 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007839 CAS Number: 719-59-5 EINECS: 211-949-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...