Dextrin, precipitated by alcohol

Supplier: Thermo Scientific Chemicals

Synonyms: Dextrin

A15717-36 A15717-22
AAA15717-36EA 123.83 USD
AAA15717-36 AAA15717-22
Dextrin, precipitated by alcohol
Dextrin

CAS No.: 9004-53-9 Merck Reference: 14,2953 MDL No.: MFCD00081554

Formula: (C₆H₁₂O₆)n
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081554
CAS Number: 9004-53-9
EINECS: 232-675-4
Merck Index: 14,2953

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR