1-Chloro-3-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloroiodobenzene

A15567-06 A15567-14 A15567-22
AAA15567-06EA 25.9 USD
AAA15567-06 AAA15567-14 AAA15567-22
1-Chloro-3-iodobenzene 98%
1-Chloro-3-iodobenzene

Purity: 98% CAS No.: 625-99-0 Molecular Formula: C6H4ClI Formula Weight: 238.46 Flash Point: 102°(216°F) Refractive Index: 1.6310 Boiling Point: 228° Beilstein Reference: 1904539 MDL No.: MFCD00001046

Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 230
Melting Pt: 54.5
Density: 1.927
Flash Pt: 102 °C (216 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001046
CAS Number: 625-99-0
EINECS: 210-920-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR