N,N,N',N',N''-Pentamethyldiethylenetriamine 98%

Supplier: Thermo Scientific Chemicals
Danger

A15433-14 A15433-36 A15433-22
AAA15433-14EA 44.8 USD
AAA15433-14 AAA15433-36 AAA15433-22
N,N,N',N',N''-Pentamethyldiethylenetriamine 98%
N,N,N',N',N''-Pentamethyldiethylenetriamine

Purity: 98% CAS No.: 3030-47-5 Molecular Formula: C9H23N3 Formula Weight: 173.30 Flash Point: 72°(161°F) Refractive Index: 1.4420 Melting Point: -20° Boiling Point: 199° Beilstein Reference: 1741396 UN No.: 2922 MDL No.: MFCD00014876

Formula: C₉H₂₃N₃
Boiling Pt: 199 °C
Melting Pt: < –20 °C
Density: 0.830
Flash Pt: 72 °C(161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014876
CAS Number: 3030-47-5
EINECS: 221-201-1
UN: 2922
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR