Benzotriazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole
A15423-18
A15423-0B
A15423-30
AAA15423-18EA
40.9
USD
AAA15423-18
AAA15423-0B
AAA15423-30
Benzotriazole 99%
Benzotriazole
Purity: 99%
CAS No.: 95-14-7
Molecular Formula: C6H5N3
Formula Weight: 119.13
Flash Point: 212°(413°F)
Physical Form: Powder
Density: 1.360
Appearance: White to pale cream
Odor: Odorless
pH: 6.0-7.0
Melting Point: 96-99°
Boiling Point: 204°/15mm
Beilstein Reference: 112133
Merck Reference: 14,1108
MDL No.: MFCD00005699
Formula:
C₆H₅N₃ MW: 119.13 g/mol Boiling Pt: 350 Melting Pt: 99 Density: 1.360 Flash Pt: 212 °C (413 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005699 CAS Number: 95-14-7 EINECS: 202-394-1 UN: 3077 ADR: 9 III Merck Index: 14,1108 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...