Picolinamide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Pyridinecarboxamide, Pyridine-2-carboxamide

A15410-14 A15410-06
AAA15410-14EA 186 USD
AAA15410-14 AAA15410-06
Picolinamide 98%
Picolinamide

Purity: 98% CAS No.: 1452-77-3 Molecular Formula: C6H6N2O Formula Weight: 122.13 Melting Point: 104-108° Boiling Point: 143°/20mm MDL No.: MFCD00023483

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 143 °C (20 mmHg)
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00023483
CAS Number: 1452-77-3
EINECS: 215-921-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR