N-Acetyl-L-cysteine 98+%
Supplier: Thermo Scientific Chemicals
A mucolytic agent for isolation of mycobacteria from sputum
Purity: 98+%
CAS No.: 616-91-1
Molecular Formula: C5H9NO3S
Formula Weight: 163.20
Melting Point: 108-111°
Beilstein Reference: 1724426
Merck Reference: 14,88
Optical Rotation: +5.3° (c=2 in water)
MDL No.: MFCD00004880
A mucolytic agent for isolation of mycobacteria from sputum
Formula:
C₅H₉NO₃S Boiling Pt: 371 °C (1013 hPa) Melting Pt: 108…111 °C Density: 1.48 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004880 CAS Number: 616-91-1 EINECS: 210-498-3 Merck Index: 14,88 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...