1,8-Octanediol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,8-Dihydroxy octane
A15402-14
A15402-22
A15402-36
AAA15402-14EA
51.8
USD
AAA15402-14
AAA15402-22
AAA15402-36
1,8-Octanediol 98+%
1,8-Octanediol
Purity: 98+%
CAS No.: 629-41-4
Molecular Formula: C8H18O2
Formula Weight: 146.23
Flash Point: 148°(298°F)
Melting Point: 58-61°
Boiling Point: 171-173°/20mm
Beilstein Reference: 1633499
MDL No.: MFCD00002989
Formula:
C₈H₁₈O₂ Boiling Pt: 171…173 °C/20 mm Melting Pt: 58…61 °C Density: 0.941 g/cm³ (20 °C) Flash Pt: 148 °C(298 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002989 CAS Number: 629-41-4 EINECS: 211-090-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...