Chloropentafluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pentafluorochlorobenzene
A15384-18
A15384-09
A15384-30
AAA15384-18EA
121
USD
AAA15384-18
AAA15384-09
AAA15384-30
Chloropentafluorobenzene 98+%
Chloropentafluorobenzene
Purity: 98+%
CAS No.: 344-07-0
Molecular Formula: C6ClF5
Formula Weight: 202.51
Refractive Index: 1.4240
Boiling Point: 116-118°
MDL No.: MFCD00000534
Formula:
C₆ClF₅ Boiling Pt: 116…118 °C Melting Pt: –15 °C Density: 1.568 Storage Temperature: Ambient |
MDL Number:
MFCD00000534 CAS Number: 344-07-0 EINECS: 206-450-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...