5-Fluoroindole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Fluoro-1H-indole

A15346-06 A15346-14 A15346-03
AAA15346-06EA 225 USD
AAA15346-06 AAA15346-14 AAA15346-03
5-Fluoroindole 99%
5-Fluoroindole

Purity: 99% CAS No.: 399-52-0 Molecular Formula: C8H6FN Formula Weight: 135.14 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 42-47° Beilstein Reference: 112350 MDL No.: MFCD00005671

Formula: C₈H₆FN
Boiling Pt: 85 °C (0.5 -rr)
Melting Pt: 42…47 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005671
CAS Number: 399-52-0
EINECS: 206-917-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR