5-Fluoroindole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Fluoro-1H-indole
A15346-06
A15346-14
A15346-03
AAA15346-06EA
225
USD
AAA15346-06
AAA15346-14
AAA15346-03
5-Fluoroindole 99%
5-Fluoroindole
Purity: 99%
CAS No.: 399-52-0
Molecular Formula: C8H6FN
Formula Weight: 135.14
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₈H₆FN Boiling Pt: 85 °C (0.5 -rr) Melting Pt: 42…47 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005671 CAS Number: 399-52-0 EINECS: 206-917-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...