Bromotrimethylsilane 97% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylbromosilane, Trimethylsilyl bromide

Stabilization: Stabilised with copper or silver

A15334-18 A15334-30 A15334-09
AAA15334-18EA 156 USD
AAA15334-18 AAA15334-30 AAA15334-09
Bromotrimethylsilane 97% stabilised
Bromotrimethylsilane

Purity: 97% CAS No.: 2857-97-8 Molecular Formula: C3H9BrSi Formula Weight: 153.09 Storage Temperature: 2°C to 8°C Flash Point: -3°(26°F) Refractive Index: 1.4250 Odor: Acrid Melting Point: -44° Boiling Point: 78-80° Beilstein Reference: 1731135 UN No.: 2924 MDL No.: MFCD00000048

Formula: C₃H₉BrSi
MW: 153.09 g/mol
Boiling Pt: 79
Melting Pt: -43
Density: 1.183
Flash Pt: –3 °C (26 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000048
CAS Number: 2857-97-8
EINECS: 220-672-0
UN: 2920
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR