Cetrimonium bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

A15235-22 A15235-36 A15235-0I
AAA15235-22EA 89.66 USD
AAA15235-22 AAA15235-36 AAAA15235-0I
Cetrimonium bromide 98%
Cetrimonium bromide

Purity: 98% CAS No.: 57-09-0 Molecular Formula: CH3(CH2)15N(CH3)3Br Formula Weight: 364.46 Flash Point: 244°(471°F) Odor: Odorless pH: 5-7 Melting Point: ca 245° dec. Beilstein Reference: 3598189 Merck Reference: 14,2025 UN No.: 3077 MDL No.: MFCD00011772

Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
Melting Pt: ∼ 245 °C (dec.)
Flash Pt: 244 °C (471 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III
Merck Index: 14,2025

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR