9-Anthracenemethanol 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
9-Anthrylmethanol, Anthracene-9-methanol
A15188-09
A15188-18
A15188-30
AAA15188-09EA
79.93
USD
AAA15188-09
AAA15188-18
AAA15188-30
9-Anthracenemethanol 97%
9-Anthracenemethanol
Purity: 97%
CAS No.: 1468-95-7
Molecular Formula: C15H12O
Formula Weight: 208.26
Melting Point: 160-165°
Beilstein Reference: 1873402
Formula:
C₁₅H₁₂O Boiling Pt: 371 °C (1013 hPa) Melting Pt: 160…165 °C Density: 1.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001264 CAS Number: 1468-95-7 EINECS: 215-998-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...