4-Chlorobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Chlorobenzoic acid, Chlorodracyclic acid, Chlorodracylic acid

A15135-0B A15135-30
AAA15135-0BEA 197 USD
AAA15135-0B AAA15135-30
4-Chlorobenzoic acid 98+%
4-Chlorobenzoic acid

Purity: 98+% CAS No.: 74-11-3 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Flash Point: 238°(460°F) Odor: Odorless Boiling Point: 275° Beilstein Reference: 907196 Merck Reference: 14,2125 MDL No.: MFCD00002531

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 287
Melting Pt: 123
Density: 1.54
Flash Pt: 238 °C (460 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002531
CAS Number: 74-11-3
EINECS: 200-805-9
Merck Index: 14,2125

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR