5-Aminoisophthalic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

A15074-30 A15074-18
AAA15074-30EA 229 USD
AAA15074-30 AAA15074-18
5-Aminoisophthalic acid 95%
5-Aminoisophthalic acid

Purity: 95% CAS No.: 99-31-0 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 300° Beilstein Reference: 2805628

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 515
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002522
CAS Number: 99-31-0
EINECS: 202-748-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR