Pivaloyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethylacetyl chloride
A15051-0F
A15051-AE
A15051-AP
AAA15051-0FEA
255
USD
AAA15051-0F
AAA15051-AE
AAA15051-AP
Pivaloyl chloride 98+%
Pivaloyl chloride
Purity: 98+%
CAS No.: 3282-30-2
Molecular Formula: (CH3)3CCOCl
Formula Weight: 120.58
Flash Point: 8°(46°F)
Refractive Index: 1.4120
Odor: Pungent
Melting Point: -57°
Boiling Point: 105-106°
Beilstein Reference: 385668
UN No.: 2438
MDL No.: MFCD00000709
Formula:
C₅H₉ClO MW: 120.58 Boiling Pt: 105 Melting Pt: -56 Density: 0.985 Flash Pt: 8 °C (46 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000709 CAS Number: 3282-30-2 EINECS: 221-921-6 UN: 2438 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...