2-Ethylaniline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14984-22
AAA14984-22EA
26.8
USD
AAA14984-22
2-Ethylaniline 97%
2-Ethylaniline
Purity: 97%
CAS No.: 578-54-1
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 91°(196°F)
Refractive Index: 1.5600
Odor: Characteristic
Melting Point: -47°
Boiling Point: 208-210°
Beilstein Reference: 1841268
UN No.: 2273
MDL No.: MFCD00007751
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 210 Melting Pt: -46 Density: 0.983 Flash Pt: 91 °C (196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007751 CAS Number: 578-54-1 EINECS: 209-424-2 UN: 2273 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...