Pyrrolidine 99%

Supplier: Thermo Scientific Chemicals
Danger

A14852-AE A14852-0F A14852-AP
AAA14852-AEEA 65.92 USD
AAA14852-AE AAA14852-0F AAA14852-AP
Pyrrolidine 99%
Pyrrolidine

MDL: MFCD00005249 Beilstein: 102395 Soluble in alcohol, ether, chloroform. Miscible with water


Purity: 99% CAS No.: 123-75-1 Molecular Formula: C4H9N Formula Weight: 71.12 Flash Point: 2°(35°F) Physical Form: Liquid Density: 0.856 Refractive Index: 1.4431 Sensitivity: Air Sensitive Appearance: Colorless to yellow Odor: Like rotten eggs Melting Point: -63° Boiling Point: 86-88° Solubility: Soluble in alcohol, ether, chloroform. Miscible with water Beilstein Reference: 102395 Merck Reference: 14,8015 UN No.: 1922 MDL No.: MFCD00005249

Formula: C₄H₉N
Boiling Pt: 86…88 °C
Melting Pt: –63 °C
Density: 0.856
Flash Pt: 2 °C (35 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005249
CAS Number: 123-75-1
EINECS: 204-648-7
UN: 1922
ADR: 3,II
Merck Index: 14,8015

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR