5-Bromo-2-furoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromofuran-2-carboxylic acid, 5-Bromofuroic acid
A14840-30
A14840-18
AAA14840-30EA
309
USD
AAA14840-30
AAA14840-18
5-Bromo-2-furoic acid 99%
5-Bromo-2-furoic acid
Purity: 99%
CAS No.: 585-70-6
Molecular Formula: C5H3BrO3
Formula Weight: 190.99
Melting Point: 188-192°
Beilstein Reference: 118354
Formula:
C₅H₃BrO₃ MW: 190.98 g/mol Melting Pt: 188…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003239 CAS Number: 585-70-6 EINECS: 209-559-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...