L-Arginine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals

A14730-36 A14730-0E A14730-22
AAA14730-36EA 214.91 USD
AAA14730-36 AAA14730-0E AAA14730-22
L-Arginine hydrochloride 98+%
L-Arginine hydrochloride

Purity: 98+% CAS No.: 1119-34-2 Molecular Formula: C6H14N4O2.HCl Formula Weight: 210.67 Melting Point: 216° dec. Beilstein Reference: 3631658 Merck Reference: 14,780 Optical Rotation: +22° (c=5 in 5N HCl) MDL No.: MFCD00064550

Formula: C₆H₁₅ClN₄O₂
Melting Pt: 216 °C (dec.)
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Merck Index: 14,780

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR