6-Aminocaproic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid

A14719-0E A14719-22 A14719-36
AAA14719-0EEA 771 USD
AAA14719-0E AAA14719-22 AAA14719-36
6-Aminocaproic acid 99%
6-Aminocaproic acid

Purity: 99% CAS No.: 60-32-2 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Physical Form: Cryst powder Appearance: White Melting Point: 210-212° Beilstein Reference: 906872 Merck Reference: 14,432 MDL No.: MFCD00008238

Formula: C₆H₁₃NO₂
Boiling Pt: 257 °C (1013 hPa)
Melting Pt: 210…212 °C
Flash Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS Number: 60-32-2
EINECS: 200-469-3
Merck Index: 14,432

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR