Manganese(II) chloride, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganous chloride, Manganese dichloride

014697-06 014697-14
AA14697-06EA 80.7 USD
AA14697-06 AAA14697-14
Manganese(II) chloride, ultra dry ≥99.998% (metals basis)
Manganese (II) chloride

MDL: MFCD00011114


Purity: 99.998% CAS No.: 7773-01-5 Molecular Formula: Cl2Mn Formula Weight: 125.84 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.977 Appearance: Pink Odor: Odorless Melting Point: 650° Boiling Point: 1190° Merck Reference: 14,5728 UN No.: 3288 MDL No.: MFCD00011114

Formula: MnCl₂
MW: 125.84 g/mol
Boiling Pt: 1190
Melting Pt: 650
Density: 2.977
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6
UN: 3288
ADR: 6.1 III
Merck Index: 14,5728

Order Now

Specification Test Results

Form -10 Mesh Beads, Ampouled under argon


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR