n-Tetradecane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99%
CAS No.: 629-59-4
Molecular Formula: C14H30
Formula Weight: 198.39
Flash Point: 99°(211°F)
Physical Form: Liquid
Density: 0.763
Refractive Index: 1.4290
Appearance: Colorless
Odor: Mild
Melting Point: 6°
Boiling Point: 252-254°
MDL No.: MFCD00008986
Formula:
H₃C(CH₂)₁₂CH₃ Boiling Pt: 252…254 °C Melting Pt: 6 °C Density: 0.763 Flash Pt: 99 °C(211 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008986 CAS Number: 629-59-4 EINECS: 211-096-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...