Phenylhydrazinium chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride

A14645-22
AAA14645-22EA 134 USD
AAA14645-22
Phenylhydrazinium chloride 99%
Phenylhydrazinium chloride

Purity: 99% CAS No.: 59-88-1 Molecular Formula: C6H8N2.HCl Formula Weight: 144.61 Melting Point: ca 252° dec. Beilstein Reference: 3594958 Merck Reference: 14,7293 UN No.: 2811 MDL No.: MFCD00012924

Formula: C₆H₉ClN₂
MW: 144.6 g/mol
Melting Pt: ∼ 252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012924
CAS Number: 59-88-1
EINECS: 200-444-7
UN: 2811
ADR: 6.1 III
Merck Index: 14,7293
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16d43c3611e20636f31c95d3974860d83c8913f&vid=c26f549d3aed1b69060ac1042fd1114ca4edb2cb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR