D-(+)-Cellobiose 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

A14553-06 A14553-14 A14553-22
AAA14553-06EA 30.6 USD
AAA14553-06 AAA14553-14 AAA14553-22
D-(+)-Cellobiose 98+%
D(+)-Cellobiose

Purity: 98+% CAS No.: 528-50-7 Molecular Formula: C12H22O11 Formula Weight: 342.30 Physical Form: Powder Appearance: White Melting Point: 234° dec. Beilstein Reference: 93795 Merck Reference: 14,1961 Optical Rotation: +34° (c=10 in water, 15h) MDL No.: MFCD00136034

Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Melting Pt: 234 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00136034
CAS Number: 528-50-7
EINECS: 208-436-5
Merck Index: 14,1961

Order Now

Specification Test Results

Optical rotation +34° (c=10 in water, 15h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR